4-Chloro-2-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16209-1G 16209-25G 16209-5G
76925-676EA 62.6 USD
76925-676 76925-678 76925-680
4-Chloro-2-fluoro-3-methoxybenzaldehyde ≥97%
4-Chloro-2-fluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19687172
CAS Number: 1002344-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR