5-Bromo-1,2-diethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

16137-1G 16137-25G 16137-500MG 16137-5G
76925-652EA 381.93 USD
76925-652 76925-654 76925-656 76925-658
5-Bromo-1,2-diethoxy-3-fluorobenzene ≥97%
5-Bromo-1,2-diethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
Storage Temperature: Refrigerator
CAS Number: 876861-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR