4-Formyl-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Formyl-2-methoxyphenyl)boronic acid

16050-25G 16050-5G
76925-636EA 566.53 USD
76925-636 76925-638
4-Formyl-2-methoxyphenylboronic acid ≥97%
4-Formyl-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856026
CAS Number: 1028479-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR