5-Bromo-2-cyanobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2-cyanophenyl)boronic acid, 5-Bromo-2-cyanophenylboronic acid

16040-25G 16040-5G
76925-626EA 488.02 USD
76925-626 76925-628
5-Bromo-2-cyanobenzeneboronic acid ≥97%
5-Bromo-2-cyanobenzeneboronic acid
Formula: C₇H₅BBrNO₂
MW: 225.836 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696674
CAS Number: 1032231-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR