4-Cyano-3,5-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16010-1G 16010-500MG 16010-5G
76925-616EA 524.09 USD
76925-616 76925-618 76925-620
4-Cyano-3,5-dimethylphenylboronic acid ≥97%
4-Cyano-3,5-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28147714
CAS Number: 911210-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR