(4-Chloro-2-fluoro-3-formylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-fluoro-3-formylphenylboronic acid

16006-1G 16006-25G 16006-500MG 16006-5G
76925-606EA 133.68 USD
76925-606 76925-608 76925-610 76925-612
(4-Chloro-2-fluoro-3-formylphenyl)boronic acid ≥97% research grade
(4-Chloro-2-fluoro-3-formylphenyl)boronic acid
Formula: C₇H₅BClFO₃
MW: 202.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617261
CAS Number: 1451393-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR