5-Fluoro-2-(hydroxymethyl)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-2-hydroxymethylphenylboronic acid, (5-Fluoro-2-(hydroxymethyl)phenyl)boronic acid

15919-1G 15919-25G 15919-5G
76925-580EA 96.55 USD
76925-580 76925-582 76925-584
5-Fluoro-2-(hydroxymethyl)benzeneboronic acid ≥97%
5-Fluoro-2-(hydroxymethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.946 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111910
CAS Number: 1246633-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR