5-Bromo-3-methyl-2-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

15839-1G 15839-25G 15839-500MG 15839-5G
76925-556EA 264.17 USD
76925-556 76925-558 76925-560 76925-562
5-Bromo-3-methyl-2-(trifluoromethyl)pyridine ≥95%
5-Bromo-3-methyl-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.021 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257678
CAS Number: 1010422-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR