5-Bromo-2-methoxyisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methoxypyridine-4-carboxaldehyde

15780-25G 15780-5G
76925-534EA 325.7 USD
76925-534 76925-536
5-Bromo-2-methoxyisonicotinaldehyde ≥97%
5-Bromo-2-methoxyisonicotinaldehyde
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856042
CAS Number: 936011-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR