4-Carboxy-2,6-dichlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15721-1G 15721-25G 15721-500MG 15721-5G
76925-520EA 231.28 USD
76925-520 76925-522 76925-524 76925-526
4-Carboxy-2,6-dichlorophenylboronic acid ≥97%
4-Carboxy-2,6-dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BCl₂O₄
MW: 234.829 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856021
CAS Number: 1451392-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR