5-Fluoroisophthalaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
15675-1G
15675-25G
15675-500MG
15675-5G
76925-498EA
144.29
USD
76925-498
76925-500
76925-502
76925-504
5-Fluoroisophthalaldehyde ≥97%
5-Fluoroisophthalaldehyde
Formula:
C₈H₅FO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08690087 CAS Number: 255375-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...