4-Ethoxy-3-fluoro-2-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15613-1G 15613-25G 15613-500MG 15613-5G
76925-486EA 224.92 USD
76925-486 76925-488 76925-490 76925-492
4-Ethoxy-3-fluoro-2-formylphenylboronic acid ≥97%
4-Ethoxy-3-fluoro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MW: 211.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856019
CAS Number: 1246404-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR