5-Bromo-3-methoxysalicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

15399-1G 15399-25G 15399-500MG 15399-5G
76925-444EA 272.66 USD
76925-444 76925-446 76925-448 76925-450
5-Bromo-3-methoxysalicylaldehyde ≥97%
5-Bromo-3-methoxysalicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00016593
CAS Number: 5034-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR