4-Methyl-3-(trifluoromethyl)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: (4-Methyl-3-(trifluoromethyl)phenyl)boronic acid, 4-Methyl-3-trifluoromethyl-phenylboronic acid, 4-Methyl-3-(trifluoromethyl)benzeneboronic acid

15264-1G 15264-25G 15264-5G
76925-434EA 108.22 USD
76925-434 76925-436 76925-438
4-Methyl-3-(trifluoromethyl)phenylboronic acid ≥95% research grade
4-Methyl-3-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₂
MW: 203.954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042431
CAS Number: 947533-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR