4-Methylthio-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15097-1G 15097-25G 15097-500MG 15097-5G
76925-408EA 201.58 USD
76925-408 76925-410 76925-412 76925-414
4-Methylthio-3-(trifluoromethyl)phenylboronic acid ≥97%
4-Methylthio-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂S
MW: 236.019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526364
CAS Number: 877383-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR