4-Chloro-2-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-trifluoromethylphenol

15048-1G 15048-25G 15048-5G
76925-398EA 66.84 USD
76925-398 76925-400 76925-402
4-Chloro-2-(trifluoromethyl)phenol ≥97%
4-Chloro-2-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
MW: 196.554 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060673
CAS Number: 53903-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR