5-Carboxy-2-chloro-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15017-1G 15017-25G 15017-500MG 15017-5G
76925-384EA 363.89 USD
76925-384 76925-386 76925-388 76925-390
5-Carboxy-2-chloro-4-fluorophenylboronic acid ≥97%
5-Carboxy-2-chloro-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClFO₄
Storage Temperature: Ambient
MDL Number: MFCD09878325
CAS Number: 957066-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR