4-Chloro-3-nitrophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-nitrobenzeneboronic acid pinacol ester, 4-Chloro-3-nitrophenylboronic acid, pinacol ester, 2-(4-Chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

14989-25G 14989-5G
76925-360EA 310.85 USD
76925-360 76925-362
4-Chloro-3-nitrophenylboronic acid pinacol ester ≥97%
4-Chloro-3-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD08235086
CAS Number: 913836-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR