2-Methyl-4-cyanophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Cyano-2-methylphenylboronic acid, (4-Cyano-2-methylphenyl)boronic acid

14872-25G 14872-5G
76925-326EA 573.95 USD
76925-326 76925-328
2-Methyl-4-cyanophenylboronic acid ≥95%
2-Methyl-4-cyanophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
MW: 160.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750473
CAS Number: 313546-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR