4-Bromo-5-fluoro-7-methyl-1H-indole ≥97%

Supplier: AOBChem USA

Warning

147854-1G 147854-500MG 147854-5G
76925-280EA 513.48 USD
76925-280 76925-282 76925-284
4-Bromo-5-fluoro-7-methyl-1H-indole ≥97%
4-Bromo-5-fluoro-7-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₇BrFN
Storage Temperature: Ambient
CAS Number: 1260895-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR