4-Bromo-5-methylthiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

14781-1G 14781-25G 14781-500MG 14781-5G
76925-270EA 296 USD
76925-270 76925-272 76925-274 76925-276
4-Bromo-5-methylthiophene-2-carboxylic acid ≥97%
4-Bromo-5-methylthiophene-2-carboxylic acid
Formula: C₆H₅BrO₂S
MW: 221.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01859873
CAS Number: 29421-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR