2-(5-Bromothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

14683-25G 14683-5G
76925-256EA 346.92 USD
76925-256 76925-258
2-(5-Bromothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(5-Bromothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₀H₁₄BBrO₂S
Storage Temperature: Ambient
MDL Number: MFCD08063136
CAS Number: 676501-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR