5-Methyl-4-bromothiophen-2-ylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-5-methylthiophene-2-boronic acid, (4-Bromo-5-methylthiophen-2-yl)boronic acid

14633-1G 14633-25G 14633-5G
76925-242EA 70.02 USD
76925-242 76925-244 76925-246
5-Methyl-4-bromothiophen-2-ylboronic acid ≥97%
5-Methyl-4-bromothiophen-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₅H₆BBrO₂S
MW: 220.88 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09952079
CAS Number: 154566-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR