4-Bromo-2-fluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

14608-1G 14608-25G 14608-500MG 14608-5G
76925-230EA 290.69 USD
76925-230 76925-232 76925-234 76925-236
4-Bromo-2-fluoro-3-methylphenol ≥97%
4-Bromo-2-fluoro-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.024 g/mol
MDL Number: MFCD28739777
CAS Number: 1807044-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR