4-Bromo-2-fluoro-1-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

14547-25G 14547-5G
76925-222EA 251.44 USD
76925-222 76925-224
4-Bromo-2-fluoro-1-propoxybenzene ≥95%
4-Bromo-2-fluoro-1-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749657
CAS Number: 127326-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR