4-Hydroxy-2-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14479-1G 14479-500MG 14479-5G
76925-210EA 456.19 USD
76925-210 76925-212 76925-214
4-Hydroxy-2-(trifluoromethoxy)phenylboronic acid ≥97%
4-Hydroxy-2-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₄
MW: 221.926 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18397563
CAS Number: 2364439-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR