5-Bromo-2-(methylthio)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methylthiopyridine, 2-Methylthio-5-bromopyridine

14365-25G 14365-5G
76925-180EA 310.85 USD
76925-180 76925-182
5-Bromo-2-(methylthio)pyridine ≥97% research grade
5-Bromo-2-(methylthio)pyridine
Formula: C₆H₆BrNS
MW: 204.0875 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411571
CAS Number: 51933-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR