Methyl-5-bromo-3-methylpicolinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester, Methyl 5-bromo-3-methylpyridine-2-carboxylate, Methyl 5-bromo-3-methylpicolinate

14283-25G 14283-5G
76925-164EA 381.93 USD
76925-164 76925-166
Methyl-5-bromo-3-methylpicolinate ≥97%
Methyl 5-bromo-3-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 230.0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375126
CAS Number: 213771-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR