5-Formyl-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

142132-1G 142132-25G 142132-500MG 142132-5G
76925-150EA 292.81 USD
76925-150 76925-152 76925-154 76925-156
5-Formyl-2,4-dimethoxybenzoic acid ≥97%
5-Formyl-2,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₅
Storage Temperature: Refrigerator
CAS Number: 633300-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR