5-Chloro-2-(trifluoromethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-Chloro-2-(trifluoromethoxy)phenyl)boronic acid

14158-1G 14158-25G 14158-5G
76925-112EA 88.06 USD
76925-112 76925-114 76925-116
5-Chloro-2-(trifluoromethoxy)phenylboronic acid ≥95%
5-Chloro-2-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₃
MW: 240.372 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697424
CAS Number: 870822-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR