3-Formyl-4-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

14138-1G 14138-25G 14138-5G
76925-094EA 84.88 USD
76925-094 76925-096 76925-098
3-Formyl-4-methoxypyridine ≥97%
3-Formyl-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD04112505
CAS Number: 82257-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR