5-Formyl-2-methoxy-4-picoline ≥97%

Supplier: AOBChem USA

Warning

14129-1G 14129-25G 14129-5G
76925-084EA 62.6 USD
76925-084 76925-086 76925-088
5-Formyl-2-methoxy-4-picoline ≥97%
5-Formyl-2-methoxy-4-picoline

Precaución: for Research Use Only

Formula: C₈H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07368879
CAS Number: 123506-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR