4-Formyl-3-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14106-1G 14106-500MG 14106-5G
76925-054EA 558.04 USD
76925-054 76925-056 76925-058
4-Formyl-3-isopropylbenzoic acid ≥97%
4-Formyl-3-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.211 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28357230
CAS Number: 1289016-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR