5-Benzyloxypyridine-3-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
14101-1G
14101-25G
14101-500MG
14101-5G
76925-038EA
251.44
USD
76925-038
76925-040
76925-042
76925-044
5-Benzyloxypyridine-3-boronic acid ≥97%
5-Benzyloxypyridine-3-boronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₂BNO₃ MW: 229.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375003 CAS Number: 1190423-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...