4-Bromo-2-isopropylaniline ≥97%

Supplier: AOBChem USA

Warning

14061-1G 14061-25G 14061-5G
76925-014EA 71.09 USD
76925-014 76925-016 76925-018
4-Bromo-2-isopropylaniline ≥97%
4-Bromo-2-isopropylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
Storage Temperature: Refrigerator
MDL Number: MFCD11845955
CAS Number: 81090-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR