5-Carboxy-2-ethoxyphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14041-1G 14041-25G 14041-500MG 14041-5G
76924-988EA 185.66 USD
76924-988 76924-990 76924-992 76924-994
5-Carboxy-2-ethoxyphenylboronic acid ≥97% research grade
5-Carboxy-2-ethoxyphenylboronic acid
Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044923
CAS Number: 1451391-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR