4-Fluoro-3-(piperidine-1-carbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14016-1G 14016-25G 14016-500MG 14016-5G
76924-952EA 124.13 USD
76924-952 76924-954 76924-956 76924-958
4-Fluoro-3-(piperidine-1-carbonyl)phenylboronic acid ≥97%
4-Fluoro-3-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235049
CAS Number: 874219-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR