5-Bromo-N-cyclopropylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

13819-1G 13819-25G 13819-5G
76924-902EA 66.84 USD
76924-902 76924-904 76924-906
5-Bromo-N-cyclopropylpicolinamide ≥97%
5-Bromo-N-cyclopropylpicolinamide

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09801062
CAS Number: 638219-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR