4,4,5,5-Tetramethyl-2-(4-methyl-3-nitrophenyl)-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Methyl-3-nitrophenylboronic acid pinacol ester

13461-25G 13461-5G
76924-876EA 559.1 USD
76924-876 76924-878
4,4,5,5-Tetramethyl-2-(4-methyl-3-nitrophenyl)-1,3,2-dioxaborolane ≥97% research grade
4,4,5,5-Tetramethyl-2-(4-methyl-3-nitrophenyl)-1,3,2-dioxaborolane
Formula: C₁₃H₁₈BNO₄
Storage Temperature: Ambient
MDL Number: MFCD09027080
CAS Number: 1072945-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR