5-Bromo-2-chloro-3-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Bromo-2-chloropyridine-3-boronic acid pinacol ester, 5-Bromo-2-chloropyridine-3-boronic acid, pinacol ester
13456-1G
13456-25G
13456-500MG
13456-5G
76924-862EA
183.54
USD
76924-862
76924-864
76924-866
76924-868
5-Bromo-2-chloro-3-pyridineboronic acid pinacol ester ≥97%
5-Bromo-2-chloro-3-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₄BBrClNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09037493 CAS Number: 1073354-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...