4-Borono-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-3-methylphenylboronic acid

13418-1G 13418-25G 13418-500MG 13418-5G
76924-846EA 189.91 USD
76924-846 76924-848 76924-850 76924-852
4-Borono-2-methylbenzoic acid ≥97%
4-Borono-2-methylbenzoic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037832
CAS Number: 191089-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR