5-Chloro-2-(pivaloylamino)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13185-1G 13185-25G 13185-500MG 13185-5G
76924-816EA 201.58 USD
76924-816 76924-818 76924-820 76924-822
5-Chloro-2-(pivaloylamino)phenylboronic acid ≥97%
5-Chloro-2-(pivaloylamino)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD01114875
CAS Number: 185950-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR