5-Bromo-2-fluoro-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

13105-25G 13105-5G
76924-810EA 587.74 USD
76924-810 76924-812
5-Bromo-2-fluoro-3-methoxypyridine ≥95%
5-Bromo-2-fluoro-3-methoxypyridine
Formula: C₆H₅BrFNO
MW: 206.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09909847
CAS Number: 880870-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR