5-Bromo-N,N-dimethylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-N,N-dimethyl-3-pyridinecarboxamide

12758-25G 12758-5G
76924-792EA 410.57 USD
76924-792 76924-794
5-Bromo-N,N-dimethylnicotinamide ≥97%
5-Bromo-N,N-dimethylnicotinamide
Formula: C₈H₉BrN₂O
MW: 229.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01788370
CAS Number: 292170-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR