(5-Bromopyridin-3-yl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

12756-25G 12756-5G
76924-784EA 187.78 USD
76924-784 76924-786
(5-Bromopyridin-3-yl)(pyrrolidin-1-yl)methanone ≥97%
(5-Bromopyridin-3-yl)(pyrrolidin-1-yl)methanone
Formula: C₁₀H₁₁BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD10000776
CAS Number: 1090388-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR