5-Bromo-3-phenyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

126050-1G 126050-25G 126050-500MG 126050-5G
76924-770EA 294.94 USD
76924-770 76924-772 76924-774 76924-776
5-Bromo-3-phenyl-1H-indazole ≥97%
5-Bromo-3-phenyl-1H-indazole
Formula: C₁₃H₉BrN₂
MW: 273.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08549635
CAS Number: 57639-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR