4-(Methylsulfonylamino)benzeneboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(Methylsulphonylamino)phenylboronic acid, 4-Methylsulphonylaminophenylboronic acid, 4-(Methanesulphonamido)phenylboronic acid

12461-1G 12461-25G 12461-5G
76924-760EA 62.6 USD
76924-760 76924-762 76924-764
4-(Methylsulfonylamino)benzeneboronic acid ≥97% research grade
4-(Methylsulfonylamino)benzeneboronic acid
Formula: C₇H₁₀BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD02179473
CAS Number: 380430-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR