4-Bromo-2-methoxy-1-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-methoxyphenyl methyl sulphone

122176-1G 122176-25G 122176-500MG 122176-5G
76924-742EA 137.92 USD
76924-742 76924-744 76924-746 76924-748
4-Bromo-2-methoxy-1-(methylsulfonyl)benzene ≥97%
4-Bromo-2-methoxy-1-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₃S
MW: 265.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22586698
CAS Number: 1095544-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR