4-Methylphenylboronic acid neopentyl glycol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methylphenylboronic acid neopentyl ester, 5,5-Dimethyl-2-(p-tolyl)-1,3,2-dioxaborinane, 4-Methylbenzeneboronic acid neopentyl glycol ester

12208-25G 12208-5G
76924-732EA 240.83 USD
76924-732 76924-734
4-Methylphenylboronic acid neopentyl glycol ester ≥97%
4-Methylphenylboronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₂
Storage Temperature: Ambient
MDL Number: MFCD03788728
CAS Number: 380481-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR