4-Methoxycarbonyl-2,6-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12172-1G 12172-25G 12172-500MG 12172-5G
76924-720EA 213.25 USD
76924-720 76924-722 76924-724 76924-726
4-Methoxycarbonyl-2,6-difluorophenylboronic acid ≥97%
4-Methoxycarbonyl-2,6-difluorophenylboronic acid
Formula: C₈H₇BF₂O₄
MW: 215.9500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095142
CAS Number: 2022984-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR